Assay Details
| Gene Symbol: S100a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761122.1 | XP_008759344.1 | 1 - 2 | 338 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008761122.1 transcripts.
Set Membership: |
|

S100a3
Genomic Map