Assay Details
| Gene Symbol: RGD1565766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239851.2 | XP_006239913.1 | 1 | 535 | 119 | |
| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009712.1 | NP_001009712.1 | 3 - 4 | 609 | 119 | |
| GenBank mRNA | BC088468.1 | - | 3 - 4 | 609 | 7127993 | 119 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1565766
Genomic Map