Assay Details
| Gene Symbol: Mina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153309.2 | NP_695221.1 | 7 - 8 | 1194 | 142 | |
| XM_008768680.1 | XP_008766902.1 | 8 - 9 | 1307 | 142 | ||
| GenBank mRNA | AB083195.1 | - | 6 - 7 | 1169 | 142 | |
| BC087650.1 | - | 7 - 8 | 1194 | 7310399 | 142 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mina
Genomic Map