Assay Details
| Gene Symbol: Ehd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024897.1 | NP_001020068.1 | 2 - 3 | 484 | 100 | |
| GenBank mRNA | BC097385.1 | - | 2 - 3 | 484 | 7446444 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Ehd2
Genomic Map