Assay Details
| Gene Symbol: Ndnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173527.1 | NP_001166998.1 | 1 | 1018 | 115 | |
| GenBank mRNA | BC098730.1 | - | 1 | 1018 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndnl2
Genomic Map