Assay Details
| Gene Symbol: Mrps31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106091.1 | NP_001099561.1 | 1 - 2 | 173 | 82 | |
| XM_006253349.2 | XP_006253411.1 | 1394 | 82 | |||
| GenBank mRNA | BC158691.1 | - | 1 - 2 | 159 | 9021908 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps31
Genomic Map