Assay Details
| Gene Symbol: C4bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012516.2 | NP_036648.2 | 9 - 10 | 1357 | 67 | |
| GenBank mRNA | BC088165.1 | - | 9 - 10 | 1357 | 7324823 | 67 |
| Z50051.1 | - | 7 - 8 | 1020 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C4bpa
Genomic Map