Assay Details
| Gene Symbol: Vegfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053653.1 | NP_446105.1 | 3 - 4 | 622 | 67 | |
| GenBank mRNA | AF010302.1 | - | 1 - 2 | 79 | 67 | |
| AY032729.1 | - | 3 - 4 | 622 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vegfc
Genomic Map