Assay Details
| Gene Symbol: Ercc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107650.2 | NP_001101120.2 | 8 - 9 | 775 | 77 | |
| NR_073139.1 | 8 - 9 | 726 | 77 | |||
| NR_073140.1 | 7 - 8 | 673 | 77 | |||
| XM_008760726.1 | XP_008758948.1 | 8 - 9 | 837 | 77 | ||
| XM_008760727.1 | XP_008758949.1 | 8 - 9 | 840 | 77 | ||
| XM_008760728.1 | XP_008758950.1 | 7 - 8 | 685 | 77 | ||
| GenBank mRNA | BC089220.1 | - | 8 - 9 | 690 | 77 | |
| BC161924.1 | - | 7 - 8 | 650 | 8370925 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ercc8
Genomic Map