Assay Details
| Gene Symbol: Sox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053349.1 | NP_445801.1 | 1 | 1853 | 146 | |
| GenBank mRNA | AJ004858.1 | - | 1 | 1853 | 146 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sox11
Genomic Map