Assay Details
| Gene Symbol: Slc39a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077669.1 | NP_001071137.1 | 3 - 4 | 761 | 66 | |
| XM_008765563.1 | XP_008763785.1 | 4 - 5 | 2938 | 66 | ||
| XM_008765564.1 | XP_008763786.1 | 4 - 5 | 2816 | 66 | ||
| GenBank mRNA | BC127514.1 | - | 3 - 4 | 761 | 7100017 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a4
Genomic Map