Assay Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107757.1 | NP_001101227.1 | 4 - 5 | 607 | 76 | |
| XM_006234707.2 | XP_006234769.1 | 5 - 6 | 981 | 76 | ||
| XM_008762093.1 | XP_008760315.1 | 5 - 6 | 981 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P192244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113149 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P192244 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P192244 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P192244 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P192244 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P192246 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88743 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10838517 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838518 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10838512 | - | - |
| Rat Genome 230 2.0 Array | 1375105_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379484_at | - | - |
| Rat Genome 230 2.0 Array | 1375105_at | - | - |
| Rat Genome 230 2.0 Array | 1379484_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375105_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379484_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379484_at | - | - |
| Affymetrix HT Rat Focus Array | 1375105_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300009501.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009506.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009502.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009504.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009507.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009496.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009503.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009505.rn.1 | TC0300001418.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300009510.rn.1 | TC0300001418.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375105_PM_at | - | - |
| Rat Expression Set 230 | 1375105_at | - | - |
| Rat Expression Set 230 | 1379484_PM_at | - | - |
| Rat Expression Set 230 | 1379484_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838512 | - | - |
| Rat Genome U34 Set | rc_AA866272_at | - | - |
| Rat Genome U34 Set | rc_AA899459_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21943035 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373780 | - | - |

Aven
Genomic Map