Assay Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114599.2 | NP_001108071.1 | 2 - 3 | 121 | 83 | |
| GenBank mRNA | BP502965.1 | - | 2 - 3 | 108 | 83 | |
| FQ213371.1 | - | 2 - 3 | 119 | 82 | ||
More Information
Set Membership: |
|

Szrd1
Genomic Map