Assay Details
| Gene Symbol: Tdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031657.1 | NP_001026827.1 | 12 - 13 | 1562 | 55 | |
| XM_006240432.2 | XP_006240494.1 | 13 - 14 | 1907 | 55 | ||
| XM_006240434.2 | XP_006240496.1 | 13 - 14 | 1578 | 55 | ||
| XM_006240435.2 | XP_006240497.1 | 13 - 14 | 2161 | 55 | ||
| GenBank mRNA | BC098739.1 | - | 12 - 13 | 1562 | 7385935 | 55 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P316755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33314 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P316755 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P316755 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P316755 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P316755 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83879 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5036 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79609 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89267 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49416 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10886354 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886351 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886356 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886353 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886358 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886355 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886352 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886357 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886349 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10886347 | - | - |
| Rat Genome 230 2.0 Array | 1377137_at | - | - |
| Rat Genome 230 2.0 Array | 1377137_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377137_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377137_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600009280.rn.1 | TC0600001341.rn.1 | RTA TC |
| Rat Expression Set 230 | 1377137_at | - | - |
| Rat Expression Set 230 | 1377137_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886347 | - | - |
| Rat Genome U34 Set | rc_AA894030_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21275240 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350291 | - | - |

Tdp1
Genomic Map