Assay Details
Gene Symbol: Tmem63c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108045.1 | NP_001101515.1 | 6 - 7 | 881 | 62 | |
XM_006240382.2 | XP_006240444.1 | 5 - 6 | 869 | 62 | ||
XM_006240383.2 | XP_006240445.1 | 7 - 8 | 1023 | 62 | ||
XM_006240384.2 | XP_006240446.1 | 6 - 7 | 654 | 62 | ||
XM_006240385.2 | XP_006240447.1 | 6 - 7 | 658 | 62 | ||
XM_006240386.2 | XP_006240448.1 | 6 - 7 | 704 | 62 | ||
XM_006240387.2 | XP_006240449.1 | 6 - 7 | 1075 | 62 | ||
XM_006240389.2 | XP_006240451.1 | 4 - 5 | 721 | 62 | ||
XM_006240390.1 | XP_006240452.1 | 1 - 2 | 55 | 62 | ||
XM_008764798.1 | XP_008763020.1 | 6 - 7 | 932 | 62 | ||
GenBank mRNA | FQ212398.1 | - | 5 - 6 | 572 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006240390.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
