Assay Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053327.1 | NP_445779.1 | 8 - 9 | 825 | 82 | |
| XM_008764291.1 | XP_008762513.1 | 8 - 9 | 941 | 82 | ||
| XM_008764292.1 | XP_008762514.1 | 8 - 9 | 911 | 82 | ||
| GenBank mRNA | BC081761.1 | - | 8 - 9 | 891 | 7108273 | 82 |
| D13927.1 | - | 8 - 9 | 825 | 82 | ||
| Z34291.1 | - | 7 - 8 | 782 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clcnka
Genomic Map