Assay Details
| Gene Symbol: Gpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153727.1 | NP_714949.1 | 1 | 901 | 76 | |
| XM_006239006.2 | XP_006239068.1 | 2 | 1037 | 76 | ||
| GenBank mRNA | AJ427482.1 | - | 1 | 901 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr3
Genomic Map