Assay Details
| Gene Symbol: Zfp444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169143.1 | NP_001162614.1 | 2 - 3 | 370 | 64 | |
| XM_006228186.2 | XP_006228248.1 | 3 - 4 | 915 | 64 | ||
| XM_008758902.1 | XP_008757124.1 | 3 - 4 | 935 | 64 | ||
| XM_008758903.1 | XP_008757125.1 | 2 - 3 | 420 | 64 | ||
| GenBank mRNA | FM030255.1 | - | 2 - 3 | 359 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp444
Genomic Map