Assay Details
| Gene Symbol: RT1-S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008886.2 | NP_001008886.2 | 5 - 6 | 1054 | 71 | |
| XM_006255917.2 | XP_006255979.1 | 5 - 6 | 1164 | 71 | ||
| GenBank mRNA | AF029241.1 | - | 5 - 6 | 979 | 71 | |
| AJ243973.1 | - | 4 - 5 | 951 | 71 | ||
| AJ243974.1 | - | 4 - 5 | 723 | 71 | ||
| AJ243975.1 | - | 4 - 5 | 951 | 71 | ||
| AJ243976.1 | - | 4 - 5 | 951 | 71 | ||
| FQ209394.1 | - | 5 - 6 | 1044 | 71 | ||
| FQ232669.1 | - | 5 - 6 | 1045 | 71 | ||
| X16979.1 | - | 4 - 5 | 774 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-S3
Genomic Map