Assay Details
| Gene Symbol: Krt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008806.1 | NP_001008806.1 | 2 - 3 | 700 | 64 | |
| XM_006242395.2 | XP_006242457.1 | 2 - 3 | 767 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krt4
Genomic Map