Assay Details
| Gene Symbol: Pcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013223.1 | NP_001013241.1 | 13 - 14 | 1119 | 80 | |
| XM_006242417.2 | XP_006242479.1 | 13 - 14 | 1132 | 80 | ||
| XM_006242418.2 | XP_006242480.1 | 13 - 14 | 1120 | 80 | ||
| XM_006242419.2 | XP_006242481.1 | 13 - 14 | 1042 | 80 | ||
| XM_006242420.2 | XP_006242482.1 | 13 - 14 | 1117 | 80 | ||
| XM_006242421.2 | XP_006242483.1 | 12 - 13 | 1093 | 80 | ||
| XM_006242422.2 | XP_006242484.1 | 12 - 13 | 1081 | 80 | ||
| XM_006242423.2 | XP_006242485.1 | 12 - 13 | 1039 | 80 | ||
| XM_006242424.2 | XP_006242486.1 | 12 - 13 | 1459 | 80 | ||
| XM_006242425.2 | XP_006242487.1 | 12 - 13 | 1027 | 80 | ||
| XM_006242426.2 | XP_006242488.1 | 11 - 12 | 1000 | 80 | ||
| XM_006242427.2 | XP_006242489.1 | 11 - 12 | 988 | 80 | ||
| XM_008765770.1 | XP_008763992.1 | 13 - 14 | 1054 | 80 | ||
| XM_008765771.1 | XP_008763993.1 | 12 - 13 | 949 | 80 | ||
| GenBank mRNA | BC078906.1 | - | 13 - 14 | 1119 | 7112287 | 80 |
| BC078909.1 | - | 12 - 13 | 949 | 7127139 | 80 | |
| BC097410.1 | - | 13 - 14 | 1144 | 7454176 | 80 | |
| FQ220695.1 | - | 8 - 9 | 607 | 80 | ||
| FQ231388.1 | - | 9 - 10 | 663 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P164575 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P024915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120178 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P024915 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164575 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P024915 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164575 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P024915 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P164575 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P371552 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3188 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97738 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47323 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86511 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27898 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49681 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51150 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75475 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36747 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74014 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84949 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72882 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42444 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17811 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94409 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57634 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76340 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27522 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49322 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69976 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39325 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10932638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932641 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899573 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899575 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932645 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899572 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914410 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899576 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10914409 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10932642 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10932636 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10899571 | - | - |
| Rat Genome 230 2.0 Array | 1398941_at | - | - |
| Rat Genome 230 2.0 Array | 1398941_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398941_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398941_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700033049.rn.1 | TC0700004040.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700033051.rn.1 | TC0700004040.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700033055.rn.1 | TC0700004040.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700033058.rn.1 | TC0700004040.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700033060.rn.1 | TC0700004040.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700033056.rn.1 | TC0700004040.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398941_PM_at | - | - |
| Rat Expression Set 230 | 1398941_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899571 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932636 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914409 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932642 | - | - |
| Rat Genome U34 Set | rc_AI104536_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21277476 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650052 | - | - |

Pcbp2
Genomic Map