Assay Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761317.1 | XP_008759539.1 | 3 - 4 | 416 | 73 | |
| XM_008761318.1 | XP_008759540.1 | 4 - 5 | 371 | 73 | ||
| XM_008761319.1 | XP_008759541.1 | 4 - 5 | 371 | 73 | ||
| XM_008761320.1 | XP_008759542.1 | 3 - 4 | 194 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008761317.1 transcripts.
Set Membership: |
|

Ciart
Genomic Map