Assay Details
| Gene Symbol: Strn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107480.2 | NP_001100950.2 | 16 - 17 | 2561 | 59 | |
| NM_001161809.1 | NP_001155281.1 | 16 - 17 | 2540 | 59 | ||
| GenBank mRNA | BC168948.1 | - | 15 - 16 | 1747 | 7308304 | 59 |
More Information
Updated Mapping Information
Set Membership: |
|

Strn4
Genomic Map