Assay Details
| Gene Symbol: Ccl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008513.1 | NP_001008513.1 | 1 - 2 | 115 | 95 | |
| GenBank mRNA | BC086571.1 | - | 1 - 2 | 115 | 7304213 | 95 |
| FQ218040.1 | - | 1 - 2 | 139 | 95 | ||
| FQ220091.1 | - | 1 - 2 | 139 | 95 | ||
| FQ223815.1 | - | 1 - 2 | 141 | 95 | ||
| FQ225478.1 | - | 1 - 2 | 139 | 95 | ||
| FQ226079.1 | - | 1 - 2 | 137 | 95 | ||
| FQ233622.1 | - | 1 - 2 | 139 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl21
Genomic Map