Assay Details
Gene Symbol: Znf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750369.3 | XP_003750417.2 | 1 - 2 | 268 | 108 | |
XM_006241919.2 | XP_006241981.1 | 1 - 2 | 268 | 108 |
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142757.1 | NP_001136229.1 | 1 - 2 | 114 | 108 | |
XM_006241927.2 | XP_006241989.1 | 1 - 2 | 268 | 108 | ||
XM_006241928.2 | XP_006241990.1 | 1 - 2 | 268 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001142757.1, XM_003750369.3, XM_006241919.2, XM_006241927.2, XM_006241928.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
