Assay Details
| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082410.1 | NP_001075879.1 | 6 - 7 | 787 | 60 | |
| XM_008764604.1 | XP_008762826.1 | 5 - 6 | 1021 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf144a
Genomic Map