Assay Details
| Gene Symbol: Creb3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007093.1 | NP_001007094.1 | 3 - 4 | 460 | 94 | |
| XM_006232627.2 | XP_006232689.1 | 4 - 5 | 518 | 94 | ||
| XM_008761147.1 | XP_008759369.1 | 3 - 4 | 615 | 94 | ||
| XM_008761148.1 | XP_008759370.1 | 3 - 4 | 690 | 94 | ||
| GenBank mRNA | AY750875.1 | - | 3 - 4 | 460 | 94 | |
| AY750876.1 | - | 3 - 4 | 454 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Creb3l4
Genomic Map