Assay Details
| Gene Symbol: Dynll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053319.3 | NP_445771.1 | 1 - 2 | 100 | 106 | |
| GenBank mRNA | BC063183.1 | - | 1 - 2 | 104 | 6887897 | 106 |
| U66461.1 | - | 1 - 2 | 87 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC063183.1, NM_053319.3, U66461.1 transcripts.
Set Membership: |
|

Dynll1
Genomic Map