Assay Details
| Gene Symbol: Ccdc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135837.1 | NP_001129309.1 | 7 | 935 | 147 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
cellular copper ion homeostasis
Golgi to plasma membrane transport
cytoplasmic sequestering of NF-kappaB
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
cullin family protein binding
cellular copper ion homeostasis
Golgi to plasma membrane transport
cytoplasmic sequestering of NF-kappaB
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
cullin family protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P785370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44320 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P785370 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P785370 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P785370 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20321 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10932543 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932545 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932540 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932555 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932554 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932549 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932556 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932553 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932544 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932541 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932546 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932550 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932552 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932551 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932548 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932542 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932547 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10932539 | - | - |
| Rat Genome 230 2.0 Array | 1372361_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372361_at | - | - |
| Affymetrix HT Rat Focus Array | 1372361_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372361_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00001175.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001180.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001191.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001179.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001193.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001195.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001183.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001174.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001173.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001181.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001194.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001196.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001184.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001186.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001188.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001177.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001185.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001190.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001192.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001198.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001176.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00001182.rn.1 | TC0X00000149.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372361_PM_at | - | - |
| Rat Expression Set 230 | 1372361_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22390056 | - | - |

Ccdc22
Genomic Map