Assay Details
| Gene Symbol: S100b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013191.1 | NP_037323.1 | 1 - 2 | 113 | 77 | |
| GenBank mRNA | BC087026.1 | - | 1 - 2 | 51 | 7193740 | 77 |
| FQ213381.1 | - | 1 - 2 | 55 | 77 | ||
| FQ213687.1 | - | 1 - 2 | 105 | 77 | ||
| FQ213774.1 | - | 1 - 2 | 107 | 77 | ||
| FQ213840.1 | - | 1 - 2 | 63 | 77 | ||
| FQ214218.1 | - | 1 - 2 | 57 | 77 | ||
| FQ214292.1 | - | 1 - 2 | 86 | 77 | ||
| FQ214554.1 | - | 1 - 2 | 55 | 77 | ||
| M54919.1 | - | 1 - 2 | 113 | 77 | ||
| X01090.1 | - | 1 - 2 | 113 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC087026.1, M54919.1, NM_013191.1, X01090.1 transcripts.
Set Membership: |
|

S100b
Genomic Map