Assay Details
| Gene Symbol: Mir702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037384.1 | 1 | 60 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | monoacylglycerol O-acyltransferase 3 | Mogat3 | -11.1kb | Sense | Chr. 12 - 22708622 - 22710784 on NCBI Build Rnor_6.0 | |
| Downstream |
zinc finger, HIT-type containing 1 |
Znhit1 | 4.7kb | Anti-sense | Chr. 12 - 22726643 - 22732917 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir702
Genomic Map