Assay Details
| Gene Symbol: LOC104845258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CF107174.1 | - | 1 - 2 | 56 | 83 | |
| CK364409.1 | - | 1 - 2 | 211 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger, DHHC-type containing 7 |
Zdhhc7 |
Serz-1, Serz1 |
-246.6kb | Sense | Chr. 19 - 52733161 - 52750307 on NCBI Build Rnor_6.0 |
| Downstream | forkhead box F1 | Foxf1 | 2.0kb | Anti-sense | Chr. 19 - 53012306 - 53016086 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

LOC104845258
Genomic Map