Assay Details
| Gene Symbol: LOC102546678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228173.2 | XP_006228235.1 | 3 - 4 | 301 | 88 | |
| XM_006228174.1 | XP_006228236.1 | 3 - 4 | 499 | 88 | ||
| XM_006228175.2 | XP_006228237.1 | 3 - 4 | 281 | 88 | ||
| XM_008758995.1 | XP_008757217.1 | 3 - 4 | 399 | 88 | ||
| XM_008758996.1 | XP_008757218.1 | 3 - 4 | 398 | 88 | ||
| Gene Symbol: Prrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253297.1 | XP_006253359.1 | 3 - 4 | 499 | 88 | |
| XM_006253298.2 | XP_006253360.1 | 3 - 4 | 281 | 88 | ||
| XM_008771339.1 | XP_008769561.1 | 3 - 4 | 399 | 88 | ||
| XM_008771340.1 | XP_008769562.1 | 3 - 4 | 398 | 88 | ||
| XM_219845.8 | XP_219845.5 | 3 - 4 | 301 | 88 | ||
More Information
Set Membership: |
|

LOC102546678
Genomic Map