Assay Details
| Gene Symbol: Pom121l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728126.4 | XP_002728172.1 | 1 | 1197 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pom121l12| Gene Symbol: Pom121l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728126.4 | XP_002728172.1 | 1 | 1197 | 98 | |
Set Membership: |
|