Assay Details
| Gene Symbol: Siglecl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759476.1 | XP_008757698.1 | 2 - 3 | 128 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008759476.1 transcripts.
Set Membership: |
|

Siglecl1
Genomic Map