Assay Details
| Gene Symbol: Uqcr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126097.2 | NP_001119569.1 | 1 - 2 | 99 | 66 | |
| NM_001287109.1 | NP_001274038.1 | 1 - 2 | 99 | 66 | ||
| XM_006241008.2 | XP_006241070.1 | 1 - 2 | 138 | 66 | ||
| GenBank mRNA | BC157815.1 | - | 1 - 2 | 75 | 8366041 | 66 |
| CB783645.1 | - | 1 - 2 | 77 | 66 | ||
| FM080527.1 | - | 1 - 2 | 101 | 66 | ||
| FQ217807.1 | - | 1 - 2 | 109 | 66 | ||
| FQ217927.1 | - | 1 - 2 | 70 | 66 | ||
More Information
Set Membership: |
|

Uqcr11
Genomic Map