Assay Details
| Gene Symbol: Car11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175708.3 | NP_783639.1 | 4 - 5 | 709 | 78 | |
| GenBank mRNA | BC086972.1 | - | 4 - 5 | 709 | 7190978 | 78 |
More Information
Set Membership: |
|

Car11
Genomic Map