Assay Details
| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009581.1 | NP_001009581.1 | 5 - 6 | 1044 | 85 | |
| GenBank mRNA | AY644721.1 | - | 5 - 6 | 1044 | 85 | |
More Information
Set Membership: |
|

ACBD3
Genomic Map