Assay Details
| Gene Symbol: IFNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111258.1 | NP_001104728.1 | 7 | 1146 | 80 | |
| GenBank mRNA | AB154365.1 | - | 7 | 1145 | 80 | |
More Information
Set Membership: |
|

IFNGR2
Genomic Map