Assay Details
| Gene Symbol: HNF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044571.1 | NP_001038036.1 | 7 - 8 | 1053 | 60 | |
| XM_005672951.3 | XP_005673008.1 | 1053 | 60 | |||
| XM_005672952.3 | XP_005673009.1 | 1052 | 60 | |||
| XM_005672953.3 | XP_005673010.1 | 922 | 60 | |||
| XM_021077331.1 | XP_020932990.1 | 922 | 60 | |||
| GenBank mRNA | AK394276.1 | - | 7 - 8 | 1033 | 60 | |
| DQ061106.1 | - | 7 - 8 | 1053 | 60 | ||
More Information
Set Membership: |
|

HNF4A
Genomic Map