Assay Details
| Gene Symbol: AMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214195.1 | NP_999360.1 | 6 - 7 | 998 | 83 | |
| XM_005663621.3 | XP_005663678.2 | 1221 | 83 | |||
| XM_005663623.3 | XP_005663680.2 | 1307 | 83 | |||
| XM_021088708.1 | XP_020944367.1 | 1303 | 83 | |||
| XM_021088709.1 | XP_020944368.1 | 1427 | 83 | |||
| XM_021088710.1 | XP_020944369.1 | 1120 | 83 | |||
| XM_021088711.1 | XP_020944370.1 | 1063 | 83 | |||
| GenBank mRNA | AF064742.1 | - | 6 - 7 | 998 | 83 | |
| AK395394.1 | - | 7 - 8 | 1159 | 83 | ||
| Gene Symbol: LOC100153854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663628.3 | XP_005663685.2 | 1104 | 83 | ||
| GenBank mRNA | AK230675.1 | - | 7 - 8 | 1618 | 83 | |
| AK395638.1 | - | 7 - 8 | 1149 | 83 | ||
| Gene Symbol: LOC100521789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125881.6 | XP_003125929.2 | 1349 | 83 | ||
| XM_005663625.3 | XP_005663682.2 | 1181 | 83 | |||
| XM_021090122.1 | XP_020945781.1 | 1287 | 83 | |||
| XM_021090123.1 | XP_020945782.1 | 1337 | 83 | |||
| XM_021090124.1 | XP_020945783.1 | 1063 | 83 | |||
| Gene Symbol: LOC110258046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081016.1 | XP_020936675.1 | 1202 | 83 | ||
| XM_021081017.1 | XP_020936676.1 | 1302 | 83 | |||
| XM_021081018.1 | XP_020936677.1 | 1184 | 83 | |||
| XM_021081019.1 | XP_020936678.1 | 1199 | 83 | |||
More Information
Set Membership: |
|

AMY2
Genomic Map