Assay Details
| Gene Symbol: MGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128482.1 | NP_001121954.1 | 1 | 1249 | 127 | |
| GenBank mRNA | AK232887.1 | - | 1 | 1696 | 127 | |
| AK233253.1 | - | 1 | 1734 | 127 | ||
| AK237196.1 | - | 3 | 1171 | 127 | ||
| AK391106.1 | - | 2 | 1504 | 127 | ||
| AK398033.1 | - | 1 | 1738 | 127 | ||
| AK398241.1 | - | 1 | 1813 | 127 | ||
More Information
Set Membership: |
|

MGAT2
Genomic Map