Assay Details
| Gene Symbol: SNRPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190213.1 | NP_001177142.1 | 2 - 3 | 249 | 78 | |
| GenBank mRNA | AK348677.1 | - | 2 - 3 | 260 | 78 | |
| BX922649.2 | - | 2 - 3 | 248 | 78 | ||
| EV898445.2 | - | 2 - 3 | 282 | 78 | ||
More Information
Set Membership: |
|

SNRPF
Genomic Map