Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244441.1 | NP_001231370.1 | 6 - 7 | 851 | 150 | |
| GenBank mRNA | AK347342.1 | - | 6 - 7 | 851 | 150 | |
| AK398387.1 | - | 4 - 5 | 731 | 150 | ||
More Information
Set Membership: |
|

FAM216A
Genomic Map