Assay Details
| Gene Symbol: CUNH14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081559.1 | XP_020937218.1 | 726 | 94 | ||
| XM_021081560.1 | XP_020937219.1 | 745 | 94 | |||
| XM_021081561.1 | XP_020937220.1 | 695 | 94 | |||
More Information
Set Membership: |
|

CUNH14orf180
Genomic Map