Assay Details
| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924465.4 | XP_001924500.1 | 393 | 79 | ||
| XM_013988209.2 | XP_013843663.1 | 364 | 79 | |||
| XM_021091498.1 | XP_020947157.1 | 200 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001924465.4, XM_021091498.1 transcripts.
Set Membership: |
|

MAFF
Genomic Map