Assay Details
| Gene Symbol: CSNK1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124088.3 | XP_003124136.1 | 1440 | 100 | ||
| XM_003354351.4 | XP_003354399.1 | 1480 | 100 | |||
| XM_005661799.2 | XP_005661856.1 | 1509 | 100 | |||
| XM_005661801.2 | XP_005661858.1 | 1490 | 100 | |||
| XM_021085007.1 | XP_020940666.1 | 1430 | 100 | |||
| GenBank mRNA | AK236633.1 | - | 2 - 3 | 148 | 100 | |
| AK236768.1 | - | 8 - 9 | 1259 | 100 | ||
| AK394472.1 | - | 4 - 5 | 383 | 100 | ||
More Information
Set Membership: |
|

CSNK1A1
Genomic Map