Assay Details
| Gene Symbol: GDPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088020.1 | XP_020943679.1 | 1259 | 61 | ||
| GenBank mRNA | AK231378.1 | - | 7 - 8 | 763 | 61 | |
| AK397276.1 | - | 7 - 8 | 1346 | 61 | ||
More Information
Set Membership: |
|

GDPD3
Genomic Map