Assay Details
| Gene Symbol: CAPSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076986.1 | XP_020932645.1 | 528 | 93 | ||
| GenBank mRNA | AK348315.1 | - | 4 - 5 | 594 | 93 | |
| AK397546.1 | - | 4 - 5 | 600 | 93 | ||
| AY609932.1 | - | 4 - 5 | 585 | 93 | ||
More Information
Set Membership: |
|

CAPSL
Genomic Map