Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128545.5 | XP_003128593.4 | 558 | 82 | ||
| XM_021099213.1 | XP_020954872.1 | 588 | 82 | |||
| GenBank mRNA | AK393910.1 | - | 2 - 3 | 235 | 82 | |
More Information
Set Membership: |
|

CHMP4A
Genomic Map